2'-O-Methylcytidine 99%

Supplier: Thermo Scientific

J65528.03EA 46.5 EUR
J65528.03
2'-O-Methylcytidine 99%
2'-O-Methylcytidine
Formula: C₁₀H₁₅N₃O₅
MW: 257,25 g/mol
MDL Number: MFCD00056067
CAS Number: 2140-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR