(S)-(+)-Tetrahydrofurfurylamine ≥98%
Supplier: Thermo Scientific
Synonyms:
(S)-(+)-2-(Aminomethyl)tetrahydrofuran
, (S)-(Tetrahydrofuran-2-yl)methanamine
L10790.03EA
0
EUR
L10790.03
(S)-(+)-Tetrahydrofurfurylamine ≥98%
(S)-(+)-Tetrahydrofurfurylamine
Formula:
C₅H₁₁NO MW: 101,15 g/mol Boiling Pt: 152…153 °C Flash Pt: 45 °C (113 °F) |
MDL Number:
MFCD00085303 CAS Number: 7175-81-7 UN: 2943 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...