Ethyl-5-chloro-1H-indole-2-carboxylate 98%

Supplier: Thermo Scientific
Warning

Synonyms: Ethyl 5-chloro-2-indolecarboxylate

L17554.03EA 32.3 EUR
L17554.03 L17554.06 L17554.14
Ethyl-5-chloro-1H-indole-2-carboxylate 98%
Ethyl-5-chloro-1H-indole-2-carboxylate
Formula: C₁₁H₁₀ClNO₂
MW: 223,66
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00005610
CAS Number: 4792-67-0
EINECS: 225-345-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR