You Searched for: "IN/Share"
Supplier:
Cayman Chemical
Description:
Potassium Phosphate Stock Solution (500 mM, pH 8.0) 1 * 250 ml
Supplier:
Cayman Chemical
Description:
Sodium Phosphate Stock Solution (500 mM, pH 7.0) 1 * 1 l
Catalog Number:
(J62982.AE)
Supplier:
Alfa Aesar
Description:
Phosphate-Citrate Buffer (5X) 1 * 100 ml
UOM:
1 * 100 ml
Catalog Number:
(HACH2307242)
Supplier:
Hach
Description:
These buffers are used in the determination of calcium and magnesium in brine.
UOM:
1 * 100 ml
Catalog Number:
(786-501)
Supplier:
G-Biosciences
Description:
A protein solubilisation buffer for 2D analysis must solubilise proteins effectively, without disturbing the native charge of the proteins. Urea based...
UOM:
1 * 50 ml
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
|
|||||||||









































