This ion selective electrode (ISE) is a half-cell with plastic body, a non-refillable internal electrolyte, and a chloride ion (Cl-) specific solid-state sensor.
- Factory controlled to ensure high quality and premium performance
- Supplied with an individual control report including information on the response curve, checkpoint value, and slope.
The ISE25Cl-9 is a half-cell ISE and requires an external reference. The Radiometer REF251, REF451, or REF601 reference electrode is recommended. As it is a screw cap electrode, it requires the CL114 cable for compatibilty with Hach sensION+ benchtop meters (or any ion meter with a BNC connection).
Delivery information: Supplied with certificate of conformity. The ISE is supplied without cable which needs to be ordered separately.