Ficoll® 400, BioChemica
Supplier: PanReac AppliChem
Ficoll®
A2252.0100EA 338 EUR
A2252.0100
Ficoll® 400, BioChemica
Cell Separation Media
Ficoll® 400 BioChemica is a non-ionic synthetic polymer of sucrose. Ficoll® 400 is a component of Denhardt's solution and several gel loading buffers. The addition of Ficoll® 400, sucrose or glycerol increase the density of the loading buffers and facilitate the loading of the sample onto the gel.
Ficoll® is a registered trademark of GE Healthcare group of companies.