You Searched For: Antibody Purification Buffers

Antibody purification buffers are necessary components in the antibody purification process, which involves selective enrichment of specific isolation of antibodies from serums, ascites fluids, or cell cultures supernatant of a hybridoma cell line. These buffers ensure high-yield, nondenaturing antibody purification, and are often provided in specialized formulations for purification of specific antibodies. Antibody purification buffers are pre-filtered and delivered ready to use, and products are available for each antibody purification method.

Antibody purification buffers are necessary components in the antibody purification process, which involves selective enrichment of specific isolation of antibodies from serums, ascites fluids, or cell cultures supernatant of a hybridoma cell line. These buffers ensure high-yield, nondenaturing antibody purification, and are often provided in specialized formulations for purification of specific antibodies. Antibody purification buffers are pre-filtered and delivered ready to use, and products are available for each antibody purification method.


8  results were found

SearchResultCount:"8"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (T6455-100ML)
Supplier: SIGMA-ALDRICH MICROSCOPY
Description: Intended for heat-induced antigen retrieval on formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies in immunohis...
UOM: 1 * 100 mL


Catalog Number: (C9999-100ML)
Supplier: SIGMA-ALDRICH MICROSCOPY
Description: Citrate buffer, antigen retriever has been used as a heat-induced antigen retriever on formalin-fixed paraffin-embedded (FFPE) tissue sections prior t...
UOM: 1 * 100 mL


Catalog Number: (ROCKDAB-B50)
Supplier: Rockland Immunochemicals
Description: Concentrate diluent buffer for DAB membrane substrate, 10X
UOM: 1 * 50 mL


Supplier: Thermo Fisher Scientific
Description: These buffers ensure high-yield, non denaturing antibody purification with IgG affinity purification supports, such as Protein A and Protein G agarose...

Catalog Number: (69755-3)
Supplier: Merck Millipore (Novagen)
Description: The His·Bind buffer kit is a set of pre-tested buffers designed for use with IDA-based His·Bind resin for convenient, rapid one-step purification of p...
UOM: 1 * 1 KIT


Catalog Number: (J63014.QA)
Supplier: Thermo Scientific
Description: Buffer kit for antibody purification
UOM: 1 * 1 KIT

Catalog Number: (28-9030-59)
Supplier: Cytiva
Description: Ab buffer kit contains prepared buffer concentrates for binding, washing, and elution of IgG according to recommended protocols.
UOM: 1 * 1 KIT


Supplier: Abcam
Description: Antibody diluent (HAMA blocker) for ELISA is a proprietary reagent for immunoassays involving a sandwich antibody complex that are susceptible to hete...

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 8 of 8
no targeter for Bottom