Detergent concentrate, PCC-54™
Supplier: Thermo Scientific
Pierce
PIER72289EA 301 EUR
PIER72289 PIER72288 PIER72290
Detergent concentrate, PCC-54™
Cleaning Agents Detergents and Surface Cleansers
PCC-54™ detergent concentrate is a general cleaning agent for use on many laboratory surface materials and glasswares including lenses, mirrors and reflectors.
  • Detergent applications include removal of dyes, blood or serum, virus and radioactive isotopes
  • Biodegradable
  • Stored at 20 to 25 °C
The cleaning detergent is mixture of anionic and nonionic surfactants. PCC-54™ detergent is strong enough to remove biological residues from plastic and glass.
A 2% solution is used for routine washing applications, or for removal of particularly stubborn dry residues then soaking in a warm 20% solution is recommended.
For removal of greases and oils then use two parts of 2% PCC-54™ detergent solution mixed with one part of ethyl alcohol.