The V-Compact is a rotational viscometer, based on the torque measurement of a rotating spindle in the sample at a specified velocity. It can be controlled via mobile devices, such as smart phones and tablets.
- Multi-language intuitive application
- Optional accessories for low ranges, small sample volumes, heldal adaptor for non flowing samples like gels and pastes
- Optional PPR system for fast and easy change of spindles
Fungilab App is an intuitive mobile application, that provides users with full functionality and control over the entire instrument. The App can be downloaded and is available free of charge.
Delivery information: Supplied with standard spindles, detector head, base, spindle protector, spindle rack, temperature probe, power supply, calibration certificate, instruction manual, carrying case.