3,5-Dibromo-1,2-phenylenediamine monohydrochloride 99%
Supplier: Thermo Scientific
Synonyms:
1,2-Diamino-3,5-dibromobenzene Monohydrochloride
, 3,5-Dibromo-o-phenylenediamine monohydrochloride
ACRO223735000EA
209
EUR
ACRO223735000
3,5-Dibromo-1,2-phenylenediamine monohydrochloride 99%
3,5-Dibromo-1,2-phenylenediamine monohydrochloride
Appearance: Beige Powder
Formula:
C₆H₆Br₂N₂ MW: 302,39 g/mol Melting Pt: 238 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012967 CAS Number: 75568-11-5 |
Specification Test Results
Appearance | Beige Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...