2-Methoxyethyl acrylate 98% stabilised

Danger

Synonyms: (2-Methoxyethyl) acrylateEthylene Glycol Monomethyl Ether AcrylateAcrylic Acid 2-Methoxyethyl EsterEthylene glycol methyl ether acrylate

ACRO335160010EA 235 EUR
ACRO335160010 ACRO335162500
2-Methoxyethyl acrylate 98% stabilised
2-Methoxyethyl acrylate

Appearance: Clear colorless to light yellow Liquid

Formula: C₆H₁₀O₃
MW: 130,14 g/mol
Boiling Pt: 56 °C (12 mmHg)
Density: 1,01 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00048149
CAS Number: 3121-61-7
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4300 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 50 to 100 ppm 4-Methoxyphenol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR