Amberlite® IRC-748 ion-exchange resin
Supplier: Thermo Scientific
ACRO335392500EA
70.7
EUR
ACRO335392500
ACRO335390010
Amberlite® IRC-748 ion-exchange resin
Amberlite® IRC-748
Appearance: Beads
Storage Temperature:
Ambient |
MDL Number:
MFCD00132702 CAS Number: 79620-28-3 |
Specification Test Results
Appearance | Beads |
Particle size |
≤1.0 % ( |
Particle size |
≤5.0 % ( |
Harmonic mean size | 0.500 to 0.650 mm |
Uniformity coefficient | ≤1.70 |
Moisture holding capacity | 60 to 65 % (Na+) |
Total volume capacity | ≥1.35 eq/l (Na+) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...