1-Benzyl-3-methylpiperidin-4-one 99%

Supplier: Thermo Scientific
Danger

Synonyms: 1-Benzyl-3-methyl-4-piperidone

ACRO134040250EA 205 EUR
ACRO134040250
1-Benzyl-3-methylpiperidin-4-one 99%
1-Benzyl-3-methylpiperidin-4-one

Appearance: Clear colorless to yellow Liquid

Formula: C₁₃H₁₇NO
MW: 203,2800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044806
CAS Number: 34737-89-8
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5315 to 1.5335 (20°C, 589 nm)
GC ≥98 %
Titration with HClO4 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR