1-(1,2,3,6-Tetrahydro-4-pyridyl)-2-benzimidazolinone 97%

Supplier: Thermo Scientific
Danger

Synonyms: 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one

ACRO135910050EA 424 EUR
ACRO135910050 ACRO135910010
1-(1,2,3,6-Tetrahydro-4-pyridyl)-2-benzimidazolinone 97%
1-(1,2,3,6-Tetrahydro-4-pyridyl)-2-benzimidazolinone

Appearance: Light yellow to beige Powder

Formula: C₁₂H₁₃N₃O
Melting Pt: 195…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00044827
CAS Number: 2147-83-3
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 195 to 200 °C
Water ≤5 % (K.F.)
HPLC ≥96.0 %
UV Lambda max 205 to 209 nm
UV Lambda max 280 to 284 nm
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR