3-Fluorosalicylaldehyde
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Fluoro-2-hydroxybenzaldehyde
ACRO398890010EA
263
EUR
ACRO398890010
3-Fluorosalicylaldehyde
3-Fluorosalicylaldehyde
Formula:
C₇H₅FO₂ MW: 140,12 g/mol Melting Pt: 68…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003319 CAS Number: 394-50-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 67 to 72 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...