Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Flame Photometers

https://ie.vwr.com/stibo/smallweb/std.lang.all/52/41/18945241.jpg

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  Cole-Parmer
Description:   The PFP7 are low temperature flame photometers designed for the determinations of sodium and potassium in industrial applications – Calcium, barium an...
Supplier:  VARIAN
Description:   The operating supplies set includes two 500 ml constant pressure bottle, a diluent bottle (1 l), a three-way SIPS tee piece assembly for SIPS-20, six ...
Supplier:  Agilent
Description:   Accessory for spectrophotometers and spectrometers, Spray cleaner for concentric nebuliser
Supplier:  VARIAN
Description:   The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
Supplier:  SHERWOOD SCIENTIFIC INSTRUMENT
Description:   Flame photometers designed for both sodium and potassium to be calibrated, measured and displayed simultaneously. The analytical performance is improv...
Supplier:  SHERWOOD SCIENTIFIC INSTRUMENT
Description:   Flame photometer is designed with front loading, unobstructed sample work area, and removable tray for easy cleaning.
Catalog Number: (SHEW47542500)

Supplier:  SHERWOOD SCIENTIFIC INSTRUMENT
Description:   Designed to measure up to four elements: Sodium (Na), potassium (K) and calcium (Ca) using air/propane flame.
UOM:  1 * 1 items
Supplier:  SHERWOOD SCIENTIFIC INSTRUMENT
Description:   Single channel flame photometer for analysis of five elements (sodium, potassium, lithium, calcium, barium) designed with front loading, unobstructed ...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 8  of 8
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping