Baskets and plate adapters, E-Z 96® Spin-Out
Supplier: Omega Bio-Tek
AC7088EA 123 EUR
OMEGAC7088
Baskets and plate adapters, E-Z 96® Spin-Out
E-Z 96® Spin-Out baskets provide a fast and convenient way to process buccal swabs, dried blood spots and other solid forensic samples in high throughput DNA extraction workflows. The unique three piece design allows for complete recovery of lysates to maximise DNA yields and also provides easier handling, which reduces overall processing time.
Samples are placed in individual basket tubes which are inserted into the E-Z® 96 Spin-Out plate adapters, which array the samples into a 96-well SBS format. The adapters can be folded in 2 different positions: one LOWERED and one RAISED. When adapters are in the LOWERED position, the basket tubes will dip into the lysis buffer and the samples are submerged, allowing for the incubation and lysis of the sample. The hinged arms of the adapters can then be folded and locked into the RAISED position, which lifts the basket tubes and samples above the liquid level. The E-Z 96® Spin-Out basket is then centrifuged to recover all the lysate within the buccal swab or other sample from the basket tubes to provide optimal DNA yields.
The E-Z 96® Spin-Out plate adapters are compatible with multiple 96-deep well, including Thermo Scientific KingFisher® 96-deep well plates and Nunc® 96-deep well plates with Shared Well Technology, thus eliminating a transfer step from 96-well plates to the plates used by the magnetic processor or manual liquid handler.