1-Cbz-piperazine 97%

Supplier: Thermo Scientific
Warning

Synonyms: Benzyl piperazine-1-carboxylatePiperazine-1-carboxylic Acid Benzyl EsterBenzyl 1-piperazinecarboxylate1-piperazinecarboxylic acid, phenylmethyl ester

ACRO367290050EA 76.9 EUR
ACRO367290050 ACRO367290250
1-Cbz-piperazine 97%
1-Cbz-piperazine
Formula: C₁₂H₁₆N₂O₂
MW: 220,272 g/mol
Boiling Pt: 158…161 °C (1,4 mmHg)
Density: 1,142 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00274317
CAS Number: 31166-44-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR