tert-Butyl-2-((diphenylmethylene)amino)acetate 98%

Supplier: Thermo Scientific

Synonyms: Glycine, N-(diphenylmethylene)-, 1,1-dimethylethyl esterN-(Diphenylmethylene)glycine tert-butyl estertert-Butyl 2-((diphenylmethylene)amino)acetate

ACRO369320010EA 121 EUR
ACRO369320010
tert-Butyl-2-((diphenylmethylene)amino)acetate 98%
tert-Butyl-2-((diphenylmethylene)amino)acetate
Formula: C₁₉H₂₁NO₂
MW: 295,382 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00134280
CAS Number: 81477-94-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 115 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR