N-Acetylguanidine 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
N-Carbamimidoylacetamide
, 1-Acetylguanidine
ACRO397440050EA
79.2
EUR
ACRO397440050
ACRO397440250
N-Acetylguanidine 97%
N-Acetylguanidine
Formula:
C₃H₇N₃O MW: 101,11 g/mol Melting Pt: 181…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075615 CAS Number: 5699-40-1 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...