2-Formylimidazole 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1H-Imidazole-2-carbaldehyde
, Imidazole-2-carboxaldehyde
ACRO398470050EA
350
EUR
ACRO398470050
ACRO398470010
2-Formylimidazole 98%
2-Formylimidazole
Formula:
C₄H₄N₂O MW: 96,09 g/mol Boiling Pt: 80 °C (,1 mmHg) Melting Pt: 209 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003544 CAS Number: 10111-08-7 |
Specification Test Results
Appearance | Off-white to light tan Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...