Dess-Martin periodinane 15% (w/w) in dichloromethane, AcroSeal®

Supplier: Thermo Scientific
Warning

Synonyms: Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl esterDMP1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one(1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one

429001000.EA 490 EUR
429001000.
Dess-Martin periodinane 15% (w/w) in dichloromethane, AcroSeal®
Dess-Martin periodinane
Formula: C₁₃H₁₃IO₈
MW: 424,15 g/mol
Density: 1,362 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00130127
CAS Number: 87413-09-0
UN: 1593
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Solution (May develop some turbidity or precipitate)
Titration Iodometric 10 to 15 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR