9-Anthraceneboronic acid 97%

Danger

Synonyms: 9-anthraceneboronic acidanthracen-9-ylboronic acidAnthracene-9-boronic acid

ACRO432810010EA 149 EUR
ACRO432810010
9-Anthraceneboronic acid 97%
9-Anthraceneboronic acid
Formula: C₁₄H₁₁BO₂
MW: 222,05 g/mol
Melting Pt: 215…218 °C
Storage Temperature: Ambient
MDL Number: MFCD03425925
CAS Number: 100622-34-2
UN: 2811

Order Now

Specification Test Results

Appearance White to tan Crystalline solid
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥99.4 %
Residue after evaporation ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR