Indole-6-carbaldehyd 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1H-Indole-6-carbaldehyde
, Indole-6-carboxaldehyde
ACRO435510010EA
290
EUR
ACRO435510010
Indole-6-carbaldehyd 97%
Indole-6-carbaldehyd
Formula:
C₉H₇NO MW: 145,16 g/mol Melting Pt: 129…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179596 CAS Number: 1196-70-9 |
Specification Test Results
Appearance | Yellow to orange to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 127 to 131 °C |
HPLC | ≥96.0 % |
Residue after evaporation | ≤0.001 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...