(R)-(-)-3-Amino-1-boc-piperidine 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(R)-1-Boc-3-Aminopiperidine
, tert-butyl (3R)-3-aminopiperidine-1-carboxylate
ACRO436490050EA
714
EUR
ACRO436490050
(R)-(-)-3-Amino-1-boc-piperidine 97%
(R)-(-)-3-Amino-1-Boc-piperidine
Formula:
C₁₀H₂₀N₂O₂ MW: 200,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094717 CAS Number: 188111-79-7 |
Specification Test Results
Appearance | Colorless to light yellow Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Specific optical rotation | -35° to -38° (20°C, 589 nm) (c=5 in MeOH) / -26.5° to -30.5° (20°C, 589 nm) (c=1, DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...