N-Fmoc-L-norleucine 98%
|
Warning
|
Synonyms:
Fmoc-Nle-OH
, FMOC-L-Norleucine
ACRO442720050EA
82.2
EUR
ACRO442720050
ACRO442720010
N-Fmoc-L-norleucine 98%
N-Fmoc-L-norleucine
Formula:
C₂₁H₂₃NO₄ MW: 353,4 g/mol Melting Pt: 138…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037537 CAS Number: 77284-32-3 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 137 to 145 °C |
HPLC | ≥97.5 % |
Specific optical rotation | -18.0° to -20.0° (20 °C, 589 nm) (c-1, DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...