6-Bromo-2-methyl-2H-indazole 97%

Supplier: Thermo Scientific
Warning

ACRO444610010EA 259 EUR
ACRO444610010
6-Bromo-2-methyl-2H-indazole 97%
6-Bromo-2-methyl-2H-indazole
Formula: C₈H₇BrN₂
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD09870048
CAS Number: 590417-95-1

Order Now

Specification Test Results

Appearance White to yellow Solid
Infrared spectrum Conforms
Melting point 83 to 88 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR