Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (2)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Formate Standards

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"4"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Formiate 1000 mg/l in water standard for ion chromatography
UOM:  1 * 100 ml
Supplier:  Alfa Aesar
Description:   Formate, Ion chromatography standard solution, Specpure|r, HCO{2}|- 1000 mg/ml 1 * 100 ml
Supplier:  ACSD
Description:   Formate standard solution, 1,000 mg/l HCO2- in water standard for ion chromatography
Supplier:  LGC Standards PROMOCHEM
Description:   This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using o...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 4  of 4
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping