Cut resistant gloves, Check & Go Red PU 1
Supplier: Honeywell Safety Products
112-4652EA 13.1 EUR
112-4652 112-4650 112-4653 112-4649
Cut resistant gloves, Check & Go Red PU 1
Gloves
These 100% nylon knitted gloves feature a red PU coating on palm and fingertips. Ideal for use when handling fine objects in a dry environment or when handling slightly greasy or dirty objects.
  • Knitting technique helps produce a glove that fits the hands snugly, with good dexterity
  • Tight knit (gauge 13) gives the glove perfect support, limits penetration of dirt and offers good sense of touch
  • Ventilated back optimises breathability
  • PU coating of the palm and fingertips provides outstanding abrasion resistance and a good grip
  • Silicone-free: Suitable for use in painting applications
  • Liner material is non pilling: Suitable for precision work
  • Colour-coding on top of the hand and coating colour helps safety manager and workers to visually recognise the level of cut protection
Typical application areas include: Administration, automotive and part manufacturing, aviation, maintenance and services.
Certifications: EN 388:2016 (3X31A); Cat. II