Daisy goblets, CBS™
Supplier: Jencons
ZZJE238-499EA 233 EUR
ZZJE238-499
Daisy goblets, CBS™
Cryogenic Straws
High security straws for freezing and storage of biological materials are placed in goblets for storage in freezers.
  • These come in eight different colours
In the Daisy configuration, goblets hold 12 visotubes (sub-compartments) of different colours and can hold a maximum of 168 straws of 0,3 or 0,5 ml.