BVIS1588
BIOV1588-1000EA 361 EUR
BIOV1588-1000 BIOV1588-100
BVIS1588
It is an EGFR kinase inhibitor.
Erlotinib is an inhibitor of human epidermal growth factor receptor (EGFR) tyrosine kinase (IC50 = 2 nM) and decreases EGFR autophosphorylation in tumor cells (IC50 = 20 nM). Studies in cell lines and enzyme assays have both shown that erlotinib inhibits EGFR at concentrations significantly lower than those needed to inhibit c-src and v-abl.