Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GABA Mouse Monoclonal Antibody [clone: AD5A9]

Supplier: MP Biomedicals
The antibody is highly specific to GABA and does not react with other amino acid-BSA conjugates with the exception of a weak reaction against b-alanine BSA-conjugate at the highest concentrations used.

  • Purified antibody is in PBS solution containing 1.0% (w/v) BSA
  • Dilution range: 1:1000 for Immunoblotting
  • Dilution range: 1:200 to 1:400 for Immunohistochemistry

Type: Primary
Antigen: GABA
Clonality: Monoclonal
Clone: AD5A9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
10 μg 10 µG ICNA08693281
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol GABA
Antigen name Gamma-Amino butyric acid
Clonality Monoclonal
Antibody type Primary
Clone AD5A9
Host Mouse
Isotype IgG1
ImmunoFluorescence Yes
Concentration 100 g/ml
Immunogen GABA - coupled to BSA with glutaraldehyde
Storage temperature 0 to +5 °C
Tested applications Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
Immunohistochemical Staining of Frozen Tissue Sections
Immunoblotting
Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
Immunohistochemical Staining of Frozen Tissue Sections
Immunoblot
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty