Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine Serum Albumin (BSA), fraction V, reagent grade

Supplier: MP Biomedicals

Albumin bovine is a single polypeptide chain consisting of about 583 amino acid residues and no carbohydrates. At pH 5 to 7 it contains 17 intrachain disulphide bridges and 1 sulphhydryl group.

  • Soluble in water
  • Has 4,7 ppm heavy metals
  • ≤5,0% loss on drying
It is used to solubilise lipids, and is also used as a blocking agent in Western blots or ELISA applications. Globulin free albumins are suitable for use in electrophoresis applications.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Bovine albumin fraction V, reagent grade 50 g ICNA08810661
Bovine albumin fraction V, reagent grade 1 kg ICNA08810667
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/peptide name Albumin
Protein synonyms Bovine Serum Albumin, BSA, Cohn Fraction V, Fraction V
Species Bovine
Source Serum
CAS No. 9048-46-8
Grade Reagent grade
Purity 100%
Storage Conditions +2 to +8 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty