Anti-IgG4 Sheep Polyclonal Antibody

Supplier: MP Biomedicals
ICNA08643131EA 317 EUR
ICNA08643131
Anti-IgG4 Sheep Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR