Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti- IgG Antibody

Supplier: MP Biomedicals
The immunoglobulin was prepared from goat serum using current chromatographic techniques. A quantitative
protein analysis as well as SDS-PAGE have been applied to determine quality of the product.

  • Supplied in Tris or phosphate buffered saline
  • Can be reconstitute with 5,0 ml of diluent for a 2% solution

It is useful for structural and biological studies, as a reference standard, or as starting material for the preparation of an immunogen.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 mg 100 mg ICNA08641431
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IgG
Antigen name Immunoglobulin G
Antibody type Secondary
Host Goat
Form Lyophilised
Storage temperature 0...5 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty