Anti-IgG Horse Antibody

Supplier: MP Biomedicals
ICNA08641441EA 207 EUR
ICNA08641441
Anti-IgG Horse Antibody
Antibodies
The immunoglobulin was prepared from horse serum using current chromatographic techniques. A quantitative
protein analysis has been applied to determine quality of the product.

  • Supplied in Tris or phosphate buffered saline
  • Can be reconstitute with 0,135 M Sodium chloride

It is useful for structural and biological studies, as a reference standard, or as starting material for the preparation of an immunogen.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope:
Host: Horse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR