Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rat Anti- IgG Antibody (Unconjugated)

Supplier: MP Biomedicals
The Immunoglobulin was isolated chromatographically from normal rat serum using DEAE and gel filtration techniques. The immunoglobulin underwent Protein G purification.

  • Can be reconstitute with 0,135 M NaCl.

It is useful for structural and biological studies, as a reference standard, or as starting material for immunogen preparation. It may be used as a reference antigen, standard, blocking agent, or coating protein in a variety of Immunoassays.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 mg 100 mg ICNA08641481
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IgG
Antigen name Immunoglobulin G
Conjugation Unconjugated
Antibody type Secondary
Host Rat
Western blot Yes
ELISA Yes
Form Liquid
Storage temperature 2...8 °C
Tested applications Immunoelectrophoresis, Immunoblot, Immunodiffusion, Dot Immunobinding
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping