Anti-BMP1 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5671-100EA 265 EUR
BIOV5671-100
Anti-BMP1 Rabbit Polyclonal Antibody
BMP-1 is an extracellular zinc endopeptidase of the Astacin family. Defects in BMP-1 are thought to lead to aberrant collagen processing and connective tissue disorders. Many forms of BMP-1 have been reported, with varying truncation at the carboxyterminal end. The long form of BMP-1 is most similar to the Tolloid-like proteins, which have extra EGF-like and CUB domains.

Type: Primary
Antigen: BMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR