Anti-IFNA2 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV6681-30TEA 101 EUR
BIOV6681-30T BIOV6681-100
Anti-IFNA2 Rabbit Polyclonal Antibody
Antibodies
Interferons (IFNs) are proteins made and released by host cells in response to the presence of pathogens. They belong to the large class of glycoproteins known as cytokines. IFN-alpha is produced by macrophages and has antiviral activities. IFNs also have other functions: they activate immune cells, such as natural killer cells and macrophages; they increase recognition of infection or tumor cells by up-regulating antigen presentation to T lymphocytes; and they increase the ability of uninfected host cells to resist new infection by virus.

Type: Primary
Antigen: IFNA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR