Anti-EPO Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV6969-100EA 299 EUR
BIOV6969-100
Anti-EPO Rabbit Polyclonal Antibody
Human erythropoietin (EPO) is an acidic glycoprotein. It is mainly produced by the kidney. As the primary regulator of the production of red cells, its major functions are to promote erythroid progenitor cells differentiation and to start the synthesis of hemoglobin. EPO acts by binding to a specific erythropoietin receptor (EPOR) on target cells; the red cell precursors in the bone marrow, and induces their transformation into mature erythrocytes. The EPO sensitivity increases with differentiation of immature progenitor cells. EPO may act as a neuron protector against glutamate toxicity. Its neuroprotective activity comes into effect by neutralizing the toxicity of free radicals. EPO is produced in brain after oxidative stress. EPO is upregulated by hypoxia and prevents apoptosis of erythroid progenitors in bone.

Type: Primary
Antigen: EPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR