Anti-Fgf2 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5039-30TEA 101 EUR
BIOV5039-30T
Anti-Fgf2 Rabbit Polyclonal Antibody
FGF-2 is a basic heparin binding growth factor that stimulates the proliferation of a wide variety of cells including mesenchymal, neuroectodermal and endothelial cells. Recombinant rat FGF-2 is a single, non-glycosylated chain containing 145 amino acids and having a molecular weight of 16.3 kDa.

Type: Primary
Antigen: Fgf2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR