Anti-FGF10 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5060A-30TEA 101 EUR
BIOV5060A-30T
Anti-FGF10 Rabbit Polyclonal Antibody
Fibroblast Growth Factor-10 (also called KGF-2) is a heparin binding growth factor that stimulates the proliferation and activation of cells that express FGF receptors. FGF-10 is mostly related to FGF-7/KGF and is expressed during development and preferentially in adult lungs.

Type: Primary
Antigen: FGF10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR