Anti-Gdnf Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5198-30TEA 101 EUR
BIOV5198-30T
Anti-Gdnf Rabbit Polyclonal Antibody
Human GDNF (Glial-Derived Neurotrophic Factor) is a 30.1 kDa protein that specifically promotes dopamine uptake and survival of midbrain neurons. GDNF is a disulphide-linked homodimer, formed by two identical 134 amino acids subunits.

Type: Primary
Antigen: Gdnf
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR