Anti-NTF3 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5306-30TEA 101 EUR
BIOV5306-30T
Anti-NTF3 Rabbit Polyclonal Antibody
Antibodies
NT-3 is a potent neurotrophic factor that supports the growth and survivability of nerve and/or glial cells. The active form of human NT-3 is a dimer, formed by two identical 119 amino acid subunits, which is held together by strong hydrophobic interactions.

Type: Primary
Antigen: NTF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR