Anti-Vegfa Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5364R-30TEA 101 EUR
BIOV5364R-30T
Anti-Vegfa Rabbit Polyclonal Antibody
Antibodies
VEGF (Vascular Endothelial Growth Factor) is a secreted homodimeric protein, secreted by a variety of vascularized tissues. VEGF stimulates endothelial cell growth, angiogenesis, and capillary permeability.

Type: Primary
Antigen: Vegfa
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR