Anti-Lep Ob Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5368-30TEA 101 EUR
BIOV5368-30T
Anti-Lep Ob Rabbit Polyclonal Antibody
Antibodies
Leptin is a recently identified protein product of the mouse obese gene. Mice with mutations in the obese gene that block the synthesis of leptin have been found to be obese and diabetic and to have reduced activity, metabolism and body temperature. cDNA clones encoding leptin have been isolated from human, simian, mouse and rat cells. The expression of leptin mRNA has been shown to be restricted to adipose tissue. Leptin plays an important role in reproduction, immunological response and neuroendocrine signaling.

Type: Primary
Antigen: Lep Ob
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR