Anti-HSP90AB1 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5858-30TEA 101 EUR
BIOV5858-30T
Anti-HSP90AB1 Rabbit Polyclonal Antibody
Antibodies
Heat shock proteins (HSPs) are ubiquitously expressed in all organisms. A major function of HSP90 and other HSPs is to act as molecular chaperones. HSP90 forms a complex with glucocorticoid receptor (GR), rendering the non-ligand-bound receptor transcriptionally inactive. HSP 90 binds the GR as a heterocomplex composed of either HSP56 or cyclophilin-40, forming an aporeceptor complex. HSP90 also exists as a dimer with other proteins such as p60/sti1 and p23, forming an aporeceptor complex with estrogen and androgen receptors.

Type: Primary
Antigen: HSP90AB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR