Anti-SELPLG Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV6657-30TEA 101 EUR
BIOV6657-30T
Anti-SELPLG Rabbit Polyclonal Antibody
Antibodies
PSGL-1 (P-Selectin glycoprotein ligand, also designated CD162), exists as a disulfide-linked homodimer. PSGL-1 is a type 1 membrane protein that localizes on the tips of microvilli of leukocytes. Its extracellular domain is rich in serines, threonines and prolines, and includes a series of 15 and 16 decameric repeats in HL-60 and U-937 cells, and human leukocytes, respectively. Although PSGL-1 appears to be the sole receptor for P-Selectin on human hematopoietic cells, it also interacts with E-Selectin through a unique binding site. In order to bind PSGL-1 to either E-Selectin or P-Selectin, PSGL-1 must be sialylated and fucosylated. PSLG-1 is a mucin-like molecule, much like leukosialin (CD43), CD164 and CD34. These proteins belong to an emerging family of cell adhesion receptors called sialomucins, which transduce negative signals in hematopoietic cells.

Type: Primary
Antigen: SELPLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR