Anti-PHF8 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV6850-25EA 209 EUR
BIOV6850-25
Anti-PHF8 Rabbit Polyclonal Antibody
PHD finger protein 8 is a Jumonji domain containing protein. Like other members of the jumonji family, PHF8 may therefore play a role in histone demethylation. Mutations in PHF8 lead to Siderius type X-linked mental retardation (MRXSSD), a mild to borderline type of mental retardation.

Type: Primary
Antigen: PHF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR