Anti-IgG Goat Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: BioVision
BIOV6907-250EA 254 EUR
BIOV6907-250
Anti-IgG Goat Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Goat Polyclonal Secondary Antibody to Rat IgG (H&L) FITC.

Type: Secondary
Antigen: IgG
Clonality: Secondary antibody
Clone:
Conjugation: Congujated
Epitope:
Host: Goat
Isotype: Rat IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR